Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGGGCATCTGATAGGGACATG[A/G]CCCTCCCCTTGGTTGAGGAGACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611254 MIM: 170290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF7 - kinesin family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLIN1 - perilipin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145311.1 | 1320 | Missense Mutation | GCC,GTC | A,V 380 | NP_001138783.1 | |
NM_002666.4 | 1320 | Missense Mutation | GCC,GTC | A,V 380 | NP_002657.3 | |
XM_005254934.4 | 1320 | Missense Mutation | GCC,GTC | A,V 380 | XP_005254991.1 |