Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTATCATTCTTTCCACTGATCCCA[A/G]AATGGCTCGATGAATGATCACAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610014 | ||||||||||||||||||||
Literature Links: |
TARSL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TARSL2 - threonyl-tRNA synthetase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152334.2 | 1522 | Silent Mutation | CTG,TTG | L,L 673 | NP_689547.2 | |
XM_006720386.3 | 1522 | Silent Mutation | CTG,TTG | L,L 673 | XP_006720449.1 | |
XM_017021912.1 | 1522 | Intron | XP_016877401.1 | |||
XM_017021913.1 | 1522 | Silent Mutation | CTG,TTG | L,L 430 | XP_016877402.1 |
TM2D3 - TM2 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |