Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGCGGCCCCTCCTGGTCTTCCT[A/G]CTGCATCCGTGCGATCAGCTGTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616844 MIM: 602437 | ||||||||||||||||||||
Literature Links: |
C15orf62 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C15orf62 - chromosome 15 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNAJC17 - DnaJ heat shock protein family (Hsp40) member C17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018163.2 | 918 | Nonsense Mutation | CAG,TAG | Q,* 296 | NP_060633.1 | |
XM_017022401.1 | 918 | Nonsense Mutation | CAG,TAG | Q,* 300 | XP_016877890.1 | |
XM_017022402.1 | 918 | Intron | XP_016877891.1 |
GCHFR - GTP cyclohydrolase I feedback regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005258.2 | 918 | Intron | NP_005249.1 |