Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCCAGCTCATAGATTGGATGGC[A/T]GTTTGGAGCGCACTGGCTCAGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605206 | ||||||||||||||||||||
Literature Links: |
HCN4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCN4 - hyperpolarization activated cyclic nucleotide gated potassium channel 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005477.2 | 4590 | Missense Mutation | CAG,CTG | Q,L 1199 | NP_005468.1 | |
XM_011521148.2 | 4590 | Missense Mutation | CAG,CTG | Q,L 793 | XP_011519450.1 |