Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCATTGATGAGGCCGCTTCCTT[C/T]CAGCACCATCGTGCAGCTGCTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGM7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGM7 - transglutaminase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052955.2 | 1939 | Missense Mutation | AAA,GAA | K,E 646 | NP_443187.1 | |
XM_017021903.1 | 1939 | Missense Mutation | AAA,GAA | K,E 647 | XP_016877392.1 |