Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGCTGATTTCTCTTTCTTTGCC[A/C]TGTCACATGTAACTCGTTTCATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603866 MIM: 170290 | ||||||||||||||||||||
Literature Links: |
PEX11A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PEX11A - peroxisomal biogenesis factor 11 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271572.1 | 494 | Missense Mutation | NP_001258501.1 | |||
NM_001271573.1 | 494 | Missense Mutation | NP_001258502.1 | |||
NM_003847.2 | 494 | Missense Mutation | NP_003838.1 |
PLIN1 - perilipin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR93 - WD repeat domain 93 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |