Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTTTTCCACAGTCAAGACATTT[A/G]TAAGGTTTCTCTCCAGTGTGGATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAL - adenosine deaminase like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBGCP4 - tubulin gamma complex associated protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN29 - zinc finger and SCAN domain containing 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152455.3 | 2705 | Silent Mutation | TAC,TAT | Y,Y 706 | NP_689668.3 | |
XM_006720401.3 | 2705 | Silent Mutation | TAC,TAT | Y,Y 706 | XP_006720464.1 | |
XM_011521266.2 | 2705 | Silent Mutation | TAC,TAT | Y,Y 706 | XP_011519568.1 | |
XM_017021949.1 | 2705 | Intron | XP_016877438.1 |