Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGAAAACACATGCTGACAGCAG[C/T]GTGTTCAGGTTTCCCTTTATTAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606087 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYNM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYNM - synemin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015286.5 | 2209 | Intron | NP_056101.5 | |||
NM_145728.2 | 2209 | Intron | NP_663780.2 | |||
XM_017022035.1 | 2209 | Intron | XP_016877524.1 |