Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAACTTCACTCACCATCTCCAC[A/G]GTGTCTGCTGGGGGTGGCCAACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102578 MIM: 608326 | ||||||||||||||||||||
Literature Links: |
PML PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PML - promyelocytic leukemia | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STOML1 - stomatin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256672.1 | 1825 | Silent Mutation | ACC,ACT | T,T 265 | NP_001243601.1 | |
NM_001256673.1 | 1825 | Silent Mutation | ACC,ACT | T,T 216 | NP_001243602.1 | |
NM_001256674.1 | 1825 | Silent Mutation | ACC,ACT | T,T 215 | NP_001243603.1 | |
NM_001256675.1 | 1825 | Intron | NP_001243604.1 | |||
NM_001256676.1 | 1825 | Silent Mutation | ACC,ACT | T,T 178 | NP_001243605.1 | |
NM_001256677.1 | 1825 | Silent Mutation | ACC,ACT | T,T 223 | NP_001243606.1 | |
NM_001324226.1 | 1825 | Silent Mutation | ACC,ACT | T,T 109 | NP_001311155.1 | |
NM_001324227.1 | 1825 | Silent Mutation | ACC,ACT | T,T 179 | NP_001311156.1 | |
NM_001324228.1 | 1825 | Silent Mutation | ACC,ACT | T,T 179 | NP_001311157.1 | |
NM_001324229.1 | 1825 | Silent Mutation | ACC,ACT | T,T 110 | NP_001311158.1 | |
NM_001324230.1 | 1825 | Silent Mutation | ACC,ACT | T,T 224 | NP_001311159.1 | |
NM_004809.4 | 1825 | Silent Mutation | ACC,ACT | T,T 266 | NP_004800.2 | |
XM_005254792.4 | 1825 | Silent Mutation | ACC,ACT | T,T 224 | XP_005254849.1 | |
XM_006720773.3 | 1825 | Silent Mutation | ACC,ACT | T,T 223 | XP_006720836.1 |