Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCAGGCACACATCTACATCCGAA[C/T]GCTCTGTGGCAGCCTCTGTAGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105370888 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105370888 - uncharacterized LOC105370888 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM202 - transmembrane protein 202 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080462.1 | 173 | Missense Mutation | ACG,ATG | T,M 58 | NP_001073931.1 | |
XM_011521497.1 | 173 | Missense Mutation | ACG,ATG | T,M 19 | XP_011519799.1 | |
XM_011521499.1 | 173 | Intron | XP_011519801.1 |