Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATAGGCTTGGTTTCTTGCTTTA[C/T]TAGGTTTATTATAAGGAGCAGTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612188 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G4F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G4F - phospholipase A2 group IVF | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS39 - VPS39, HOPS complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301138.1 | 4905 | UTR 3 | NP_001288067.1 | |||
NM_015289.3 | 4905 | UTR 3 | NP_056104.2 | |||
XM_011521403.2 | 4905 | UTR 3 | XP_011519705.1 | |||
XM_011521404.2 | 4905 | UTR 3 | XP_011519706.1 | |||
XM_017022036.1 | 4905 | Intron | XP_016877525.1 | |||
XM_017022037.1 | 4905 | Intron | XP_016877526.1 | |||
XM_017022038.1 | 4905 | UTR 3 | XP_016877527.1 | |||
XM_017022039.1 | 4905 | UTR 3 | XP_016877528.1 |