Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTGGCCAATGACTCGCAACCACT[C/T]ACCGTCTACTTCTCCCTGAGCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRNA7 - cholinergic receptor nicotinic alpha 7 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000746.5 | 203 | Silent Mutation | CTC,CTT | L,L 51 | NP_000737.1 | |
NM_001190455.2 | 203 | Silent Mutation | CTC,CTT | L,L 80 | NP_001177384.1 | |
XM_011521176.2 | 203 | Intron | XP_011519478.2 | |||
XM_011521177.2 | 203 | Intron | XP_011519479.1 | |||
XM_011521178.2 | 203 | Silent Mutation | CTC,CTT | L,L 51 | XP_011519480.1 | |
XM_017021882.1 | 203 | UTR 5 | XP_016877371.1 | |||
XM_017021883.1 | 203 | UTR 5 | XP_016877372.1 | |||
XM_017021884.1 | 203 | UTR 5 | XP_016877373.1 |
LOC105370940 - uncharacterized LOC105370940 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011522320.2 | 203 | Intron | XP_011520622.1 |