Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGCCTGGCCCCCGGCTCGCCCT[A/G]TGTTTCGTGGACATCCGCCGGTTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616701 MIM: 154580 MIM: 608844 | ||||||||||||||||||||
Literature Links: |
COMMD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COMMD4 - COMM domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAN2C1 - mannosidase alpha class 2C member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR631 - microRNA 631 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEIL1 - nei like DNA glycosylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256552.1 | 761 | Silent Mutation | CTA,CTG | L,L 198 | NP_001243481.1 | |
NM_024608.3 | 761 | Silent Mutation | CTA,CTG | L,L 112 | NP_078884.2 | |
XM_005254659.4 | 761 | Silent Mutation | CTA,CTG | L,L 112 | XP_005254716.1 | |
XM_006720677.3 | 761 | Silent Mutation | CTA,CTG | L,L 189 | XP_006720740.1 | |
XM_006720678.3 | 761 | Silent Mutation | CTA,CTG | L,L 198 | XP_006720741.1 | |
XM_006720679.3 | 761 | Silent Mutation | CTA,CTG | L,L 155 | XP_006720742.1 | |
XM_006720680.1 | 761 | Silent Mutation | CTA,CTG | L,L 112 | XP_006720743.1 | |
XM_006720681.1 | 761 | Silent Mutation | CTA,CTG | L,L 112 | XP_006720744.1 | |
XM_011522001.2 | 761 | Silent Mutation | CTA,CTG | L,L 175 | XP_011520303.2 | |
XM_011522002.1 | 761 | Silent Mutation | CTA,CTG | L,L 112 | XP_011520304.1 | |
XM_011522003.2 | 761 | Silent Mutation | CTA,CTG | L,L 112 | XP_011520305.1 | |
XM_011522004.2 | 761 | Silent Mutation | CTA,CTG | L,L 112 | XP_011520306.1 | |
XM_011522005.1 | 761 | Missense Mutation | ATG,GTG | M,V 1 | XP_011520307.1 | |
XM_011522006.1 | 761 | Missense Mutation | TAT,TGT | Y,C 17 | XP_011520308.1 | |
XM_011522007.1 | 761 | Missense Mutation | TAT,TGT | Y,C 17 | XP_011520309.1 | |
XM_011522008.1 | 761 | Missense Mutation | TAT,TGT | Y,C 17 | XP_011520310.1 | |
XM_017022566.1 | 761 | Silent Mutation | CTA,CTG | L,L 189 | XP_016878055.1 | |
XM_017022567.1 | 761 | Missense Mutation | TAT,TGT | Y,C 17 | XP_016878056.1 | |
XM_017022568.1 | 761 | Silent Mutation | CTA,CTG | L,L 198 | XP_016878057.1 |