Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCCCAAAGCATGGAGGATTTT[A/G]TAGATCTCCTTGATGTTCAACCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191316 | ||||||||||||||||||||
Literature Links: |
NTRK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NTRK3 - neurotrophic receptor tyrosine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007156.2 | 2388 | Intron | NP_001007157.1 | |||
NM_001012338.2 | 2388 | Silent Mutation | TAC,TAT | Y,Y 821 | NP_001012338.1 | |
NM_001243101.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 799 | NP_001230030.1 | |
NM_001320134.1 | 2388 | Intron | NP_001307063.1 | |||
NM_001320135.1 | 2388 | Intron | NP_001307064.1 | |||
NM_002530.3 | 2388 | Silent Mutation | TAC,TAT | Y,Y 807 | NP_002521.2 | |
XM_006720543.3 | 2388 | Silent Mutation | TAC,TAT | Y,Y 807 | XP_006720606.1 | |
XM_006720544.3 | 2388 | Silent Mutation | TAC,TAT | Y,Y 799 | XP_006720607.1 | |
XM_006720545.3 | 2388 | Intron | XP_006720608.1 | |||
XM_006720548.3 | 2388 | Intron | XP_006720611.1 | |||
XM_006720549.3 | 2388 | Intron | XP_006720612.1 | |||
XM_006720550.3 | 2388 | Intron | XP_006720613.1 | |||
XM_011521637.2 | 2388 | Intron | XP_011519939.1 | |||
XM_011521638.2 | 2388 | Intron | XP_011519940.1 | |||
XM_017022240.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 807 | XP_016877729.1 | |
XM_017022241.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 745 | XP_016877730.1 | |
XM_017022242.1 | 2388 | Intron | XP_016877731.1 | |||
XM_017022243.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 709 | XP_016877732.1 | |
XM_017022244.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 709 | XP_016877733.1 | |
XM_017022245.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 709 | XP_016877734.1 | |
XM_017022246.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 709 | XP_016877735.1 | |
XM_017022247.1 | 2388 | Intron | XP_016877736.1 | |||
XM_017022248.1 | 2388 | Intron | XP_016877737.1 | |||
XM_017022249.1 | 2388 | Intron | XP_016877738.1 | |||
XM_017022250.1 | 2388 | Intron | XP_016877739.1 | |||
XM_017022251.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 438 | XP_016877740.1 | |
XM_017022252.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 430 | XP_016877741.1 | |
XM_017022253.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 821 | XP_016877742.1 | |
XM_017022254.1 | 2388 | Silent Mutation | TAC,TAT | Y,Y 832 | XP_016877743.1 |