Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATGTGCTGCAAATGTCTTCTGT[C/T]GGTTTGTAGCCTGTCTCTTCACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARP6 - poly(ADP-ribose) polymerase family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323515.1 | Intron | NP_001310444.1 | ||||
NM_001323516.1 | Intron | NP_001310445.1 | ||||
NM_001323519.1 | Intron | NP_001310448.1 | ||||
NM_001323521.1 | Intron | NP_001310450.1 | ||||
NM_001323522.1 | Intron | NP_001310451.1 | ||||
NM_001323523.1 | Intron | NP_001310452.1 | ||||
NM_001323524.1 | Intron | NP_001310453.1 | ||||
NM_001323525.1 | Intron | NP_001310454.1 | ||||
NM_001323526.1 | Intron | NP_001310455.1 | ||||
NM_001323528.1 | Intron | NP_001310457.1 | ||||
NM_001323530.1 | Intron | NP_001310459.1 | ||||
NM_001323531.1 | Intron | NP_001310460.1 | ||||
NM_001323532.1 | Intron | NP_001310461.1 | ||||
NM_020214.3 | Intron | NP_064599.2 | ||||
XM_011521805.1 | Intron | XP_011520107.1 | ||||
XM_017022418.1 | Intron | XP_016877907.1 |