Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGATCTCCCAGAGCAGCACCCCA[A/G]AAGACCTGCATCACAAGTGGGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147521 MIM: 151520 | ||||||||||||||||||||
Literature Links: |
ITPKA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITPKA - inositol-trisphosphate 3-kinase A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTK - leukocyte receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135685.1 | 2223 | Missense Mutation | TCT,TTT | S,F 579 | NP_001129157.1 | |
NM_002344.5 | 2223 | Missense Mutation | TCT,TTT | S,F 709 | NP_002335.2 | |
NM_206961.3 | 2223 | Missense Mutation | TCT,TTT | S,F 648 | NP_996844.1 | |
XM_011521556.2 | 2223 | Missense Mutation | TCT,TTT | S,F 725 | XP_011519858.1 | |
XM_011521557.2 | 2223 | Missense Mutation | TCT,TTT | S,F 669 | XP_011519859.1 | |
XM_017022181.1 | 2223 | Missense Mutation | TCT,TTT | S,F 730 | XP_016877670.1 | |
XM_017022182.1 | 2223 | Missense Mutation | TCT,TTT | S,F 682 | XP_016877671.1 | |
XM_017022183.1 | 2223 | Missense Mutation | TCT,TTT | S,F 645 | XP_016877672.1 |