Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAACTCATTTTGAGAAAGAAGCT[G/T]CTCTTTGTGGCCAAACACACCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615276 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CERS3 - ceramide synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290341.2 | 1990 | UTR 3 | NP_001277270.1 | |||
NM_001290342.2 | 1990 | UTR 3 | NP_001277271.1 | |||
NM_001290343.1 | 1990 | UTR 3 | NP_001277272.1 | |||
NM_178842.4 | 1990 | UTR 3 | NP_849164.2 | |||
XM_011521355.2 | 1990 | UTR 3 | XP_011519657.1 | |||
XM_011521357.2 | 1990 | UTR 3 | XP_011519659.1 | |||
XM_011521358.1 | 1990 | UTR 3 | XP_011519660.1 | |||
XM_017022002.1 | 1990 | UTR 3 | XP_016877491.1 | |||
XM_017022003.1 | 1990 | UTR 3 | XP_016877492.1 | |||
XM_017022004.1 | 1990 | UTR 3 | XP_016877493.1 |
CERS3-AS1 - CERS3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |