Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCGACTGAGGAGCTGGCCCCC[A/G]GTGGCCTGCACCGTCCAGCGGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610953 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIF1 - PIF1 5'-to-3' DNA helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286496.1 | 1645 | Silent Mutation | NP_001273425.1 | |||
NM_001286497.1 | 1645 | Silent Mutation | NP_001273426.1 | |||
NM_001286499.1 | 1645 | Silent Mutation | NP_001273428.1 | |||
NM_025049.3 | 1645 | Silent Mutation | NP_079325.2 | |||
XM_011522083.2 | 1645 | Silent Mutation | XP_011520385.1 | |||
XM_011522084.2 | 1645 | Silent Mutation | XP_011520386.1 |