Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTTCTGCTGAACCTGTGGTAA[A/C]ATATTCCTCATAGGGATCCAAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612186 | ||||||||||||||||||||
Literature Links: |
DMXL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DMXL2 - Dmx like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174116.1 | 9220 | Missense Mutation | GTT,TTT | V,F 2958 | NP_001167587.1 | |
NM_001174117.1 | 9220 | Missense Mutation | GTT,TTT | V,F 2321 | NP_001167588.1 | |
NM_015263.3 | 9220 | Missense Mutation | GTT,TTT | V,F 2957 | NP_056078.2 | |
XM_005254255.1 | 9220 | Missense Mutation | GTT,TTT | V,F 2979 | XP_005254312.1 | |
XM_005254256.1 | 9220 | Missense Mutation | GTT,TTT | V,F 2978 | XP_005254313.1 | |
XM_011521402.2 | 9220 | Intron | XP_011519704.1 | |||
XM_017022034.1 | 9220 | Missense Mutation | GTT,TTT | V,F 2961 | XP_016877523.1 |