Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGGAGATACTTAAATATAGACAT[C/T]GAATCTTGCAATTTCAAAATCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC91450 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC91450 - uncharacterized LOC91450 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D2B - TBC1 domain family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015079.5 | 2643 | Silent Mutation | TCA,TCG | S,S 881 | NP_055894.6 | |
NM_144572.1 | 2643 | Silent Mutation | TCA,TCG | S,S 881 | NP_653173.1 | |
XM_011521387.2 | 2643 | Silent Mutation | TCA,TCG | S,S 844 | XP_011519689.1 | |
XM_017022022.1 | 2643 | Silent Mutation | TCA,TCG | S,S 651 | XP_016877511.1 |