Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAGTGGGCAATCTAGTAGGCCA[A/G]TTCACATTACAGTTATCTCAGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609096 MIM: 610894 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO22 - F-box protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012170.3 | 4079 | Intron | NP_036302.1 | |||
NM_147188.2 | 4079 | Intron | NP_671717.1 | |||
XM_011521447.2 | 4079 | UTR 3 | XP_011519749.1 |
FBXO22-AS1 - FBXO22 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRG4 - neuregulin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138573.3 | 4079 | Intron | NP_612640.1 | |||
XM_017021937.1 | 4079 | UTR 3 | XP_016877426.1 | |||
XM_017021938.1 | 4079 | UTR 3 | XP_016877427.1 | |||
XM_017021939.1 | 4079 | UTR 3 | XP_016877428.1 | |||
XM_017021940.1 | 4079 | UTR 3 | XP_016877429.1 | |||
XM_017021941.1 | 4079 | UTR 3 | XP_016877430.1 | |||
XM_017021942.1 | 4079 | UTR 3 | XP_016877431.1 | |||
XM_017021943.1 | 4079 | UTR 3 | XP_016877432.1 | |||
XM_017021944.1 | 4079 | UTR 3 | XP_016877433.1 | |||
XM_017021945.1 | 4079 | Intron | XP_016877434.1 | |||
XM_017021946.1 | 4079 | UTR 3 | XP_016877435.1 | |||
XM_017021947.1 | 4079 | Intron | XP_016877436.1 | |||
XM_017021948.1 | 4079 | Intron | XP_016877437.1 |