Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATAAGGCACAGCCCCGGCAGGGA[C/T]TTAATGCTGGCAAGTGCAGGCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G4E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G4E - phospholipase A2 group IVE | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206670.1 | Intron | NP_001193599.1 | ||||
XM_011521237.2 | Intron | XP_011519539.1 | ||||
XM_011521238.1 | Intron | XP_011519540.1 | ||||
XM_011521239.1 | Intron | XP_011519541.1 |
PLA2G4E-AS1 - PLA2G4E antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |