Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAATCACAAATCACCACAAAAG[C/T]AGTGGCTTAAAGCAACACAAGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAQR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAQR5 - progestin and adipoQ receptor family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104554.1 | Intron | NP_001098024.1 | ||||
NM_017705.3 | Intron | NP_060175.3 | ||||
XM_005254495.4 | Intron | XP_005254552.1 | ||||
XM_011521720.2 | Intron | XP_011520022.1 | ||||
XM_011521722.2 | Intron | XP_011520024.1 | ||||
XM_011521723.2 | Intron | XP_011520025.1 | ||||
XM_011521725.2 | Intron | XP_011520027.1 | ||||
XM_011521727.2 | Intron | XP_011520029.1 | ||||
XM_017022360.1 | Intron | XP_016877849.1 | ||||
XM_017022361.1 | Intron | XP_016877850.1 | ||||
XM_017022362.1 | Intron | XP_016877851.1 | ||||
XM_017022363.1 | Intron | XP_016877852.1 | ||||
XM_017022364.1 | Intron | XP_016877853.1 | ||||
XM_017022365.1 | Intron | XP_016877854.1 | ||||
XM_017022366.1 | Intron | XP_016877855.1 | ||||
XM_017022367.1 | Intron | XP_016877856.1 | ||||
XM_017022368.1 | Intron | XP_016877857.1 |