Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACTGAAGCAGGCCCAGACCCTG[A/C]TGAGCTCCCTGGGCTGAGGAGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616701 MIM: 608844 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COMMD4 - COMM domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284377.1 | 655 | Missense Mutation | ATG,CTG | M,L 136 | NP_001271306.1 | |
NM_001284378.1 | 655 | Missense Mutation | ATG,CTG | M,L 109 | NP_001271307.1 | |
NM_001284379.1 | 655 | Missense Mutation | ATG,CTG | M,L 142 | NP_001271308.1 | |
NM_001321844.1 | 655 | Missense Mutation | ATG,CTG | M,L 171 | NP_001308773.1 | |
NM_001321845.1 | 655 | Missense Mutation | ATG,CTG | M,L 149 | NP_001308774.1 | |
NM_001321846.1 | 655 | Missense Mutation | ATG,CTG | M,L 114 | NP_001308775.1 | |
NM_001321847.1 | 655 | Missense Mutation | ATG,CTG | M,L 173 | NP_001308776.1 | |
NM_001321848.1 | 655 | Missense Mutation | ATG,CTG | M,L 201 | NP_001308777.1 | |
NM_017828.4 | 655 | Missense Mutation | ATG,CTG | M,L 195 | NP_060298.2 | |
XM_005254509.3 | 655 | Missense Mutation | ATG,CTG | M,L 222 | XP_005254566.1 | |
XM_005254510.3 | 655 | Missense Mutation | ATG,CTG | M,L 200 | XP_005254567.1 | |
XM_011521739.1 | 655 | Missense Mutation | ATG,CTG | M,L 225 | XP_011520041.1 | |
XM_011521740.1 | 655 | Missense Mutation | ATG,CTG | M,L 203 | XP_011520042.1 | |
XM_011521741.1 | 655 | Missense Mutation | ATG,CTG | M,L 166 | XP_011520043.1 | |
XM_017022385.1 | 655 | Missense Mutation | ATG,CTG | M,L 144 | XP_016877874.1 |
NEIL1 - nei like DNA glycosylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |