Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGGGCCTCTACAGCTTCAGCA[G/T]CAGCCCAGATAAAGCCCAGCCAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123841 | ||||||||||||||||||||
Literature Links: |
PPIB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPIB - peptidylprolyl isomerase B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX22 - sorting nexin 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024798.2 | 577 | Missense Mutation | AGC,ATC | S,I 173 | NP_079074.2 | |
XM_005254677.3 | 577 | Intron | XP_005254734.1 | |||
XM_017022581.1 | 577 | Missense Mutation | AGC,ATC | S,I 145 | XP_016878070.1 |