Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGAGCTCACCTTCGAGCTGCC[G/T]GACAACGCCAAGCAGTGCTTCCACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMED3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMED3 - transmembrane p24 trafficking protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301203.1 | 290 | Silent Mutation | CCG,CCT | P,P 34 | NP_001288132.1 | |
NM_007364.3 | 290 | Silent Mutation | CCG,CCT | P,P 34 | NP_031390.1 | |
XM_017022040.1 | 290 | Silent Mutation | CCG,CCT | P,P 34 | XP_016877529.1 | |
XM_017022041.1 | 290 | Silent Mutation | CCG,CCT | P,P 34 | XP_016877530.1 |