Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGGAGGGCAGCGCTCCACGCC[C/T]CTCATCATCGCAGCCCGCAATGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613539 | ||||||||||||||||||||
Literature Links: |
FEM1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FEM1B - fem-1 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015322.4 | 762 | Silent Mutation | CCC,CCT | P,P 49 | NP_056137.1 | |
XM_017021859.1 | 762 | Intron | XP_016877348.1 |