Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGATTGGAAATTCATAGCCCAG[A/G]TTCTTGATCGGATGTTTCTGTGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118503 MIM: 118505 | ||||||||||||||||||||
Literature Links: |
CHRNA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNA3 - cholinergic receptor nicotinic alpha 3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000743.4 | 1960 | Intron | NP_000734.2 | |||
NM_001166694.1 | 1960 | Missense Mutation | NP_001160166.1 | |||
XM_006720382.2 | 1960 | Intron | XP_006720445.1 |
CHRNA5 - cholinergic receptor nicotinic alpha 5 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000745.3 | 1960 | Missense Mutation | ATT,GTT | I,V 426 | NP_000736.2 | |
NM_001307945.1 | 1960 | UTR 3 | NP_001294874.1 | |||
XM_005254142.3 | 1960 | UTR 3 | XP_005254199.1 | |||
XM_017021881.1 | 1960 | UTR 3 | XP_016877370.1 |