Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAGCTAAGGCTGCAAATTAAA[C/T]CACTACCACACAGCTGCAAACTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLTM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLTM - SAFB like transcription modulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013843.2 | 3324 | UTR 3 | NP_001013865.1 | |||
NM_024755.3 | 3324 | UTR 3 | NP_079031.2 | |||
XM_006720686.3 | 3324 | UTR 3 | XP_006720749.3 | |||
XM_006720690.1 | 3324 | UTR 3 | XP_006720753.1 | |||
XM_011522022.1 | 3324 | UTR 3 | XP_011520324.1 | |||
XM_011522023.1 | 3324 | UTR 3 | XP_011520325.1 | |||
XM_011522024.1 | 3324 | UTR 3 | XP_011520326.1 | |||
XM_011522025.1 | 3324 | UTR 3 | XP_011520327.1 | |||
XM_011522026.2 | 3324 | UTR 3 | XP_011520328.2 | |||
XM_011522027.1 | 3324 | UTR 3 | XP_011520329.1 | |||
XM_011522028.1 | 3324 | UTR 3 | XP_011520330.1 | |||
XM_011522029.2 | 3324 | UTR 3 | XP_011520331.1 | |||
XM_011522030.2 | 3324 | Intron | XP_011520332.1 | |||
XM_011522031.2 | 3324 | UTR 3 | XP_011520333.1 | |||
XM_017022576.1 | 3324 | UTR 3 | XP_016878065.1 | |||
XM_017022577.1 | 3324 | UTR 3 | XP_016878066.1 | |||
XM_017022578.1 | 3324 | UTR 3 | XP_016878067.1 | |||
XM_017022579.1 | 3324 | UTR 3 | XP_016878068.1 | |||
XM_017022580.1 | 3324 | UTR 3 | XP_016878069.1 |