Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACAGGGGAATGTCCTGGGACTT[C/G]GGACTGTCAGCCATGGACCGGTAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606759 MIM: 612771 MIM: 612772 | ||||||||||||||||||||
Literature Links: |
DUOX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUOX2 - dual oxidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUOXA1 - dual oxidase maturation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276264.1 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | NP_001263193.1 | |
NM_001276265.1 | 1390 | Silent Mutation | CCC,CCG | P,P 269 | NP_001263194.1 | |
NM_001276266.1 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | NP_001263195.1 | |
NM_001276267.1 | 1390 | Silent Mutation | CCC,CCG | P,P 269 | NP_001263196.1 | |
NM_001276268.1 | 1390 | Silent Mutation | CCC,CCG | P,P 269 | NP_001263197.1 | |
NM_144565.3 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | NP_653166.2 | |
XM_006720743.3 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_006720806.1 | |
XM_006720745.1 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_006720808.1 | |
XM_006720746.1 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_006720809.1 | |
XM_006720747.3 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_006720810.1 | |
XM_006720749.1 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_006720812.1 | |
XM_006720751.3 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_006720814.1 | |
XM_006720752.3 | 1390 | Silent Mutation | CCC,CCG | P,P 343 | XP_006720815.2 | |
XM_006720754.1 | 1390 | Silent Mutation | CCC,CCG | P,P 269 | XP_006720817.1 | |
XM_006720755.1 | 1390 | Silent Mutation | CCC,CCG | P,P 269 | XP_006720818.1 | |
XM_011522181.2 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_011520483.1 | |
XM_011522182.1 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_011520484.1 | |
XM_011522183.2 | 1390 | Silent Mutation | CCC,CCG | P,P 314 | XP_011520485.1 | |
XM_011522184.1 | 1390 | Intron | XP_011520486.1 | |||
XM_011522185.1 | 1390 | UTR 3 | XP_011520487.1 | |||
XM_011522186.2 | 1390 | Intron | XP_011520488.1 |
DUOXA2 - dual oxidase maturation factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |