Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCTCGATACCTGTTTTAACAGG[C/G]TGATGTCCAAGTGCAGTTGATCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180479 MIM: 609984 | ||||||||||||||||||||
Literature Links: |
RPL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPL4 - ribosomal protein L4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZWILCH - zwilch kinetochore protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287821.1 | 458 | UTR 5 | NP_001274750.1 | |||
NM_001287822.1 | 458 | Intron | NP_001274751.1 | |||
NM_001287823.1 | 458 | UTR 5 | NP_001274752.1 | |||
NM_017975.4 | 458 | Missense Mutation | GCT,GGT | A,G 36 | NP_060445.3 |