Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTTATGGAGCCAGCTGGCTGC[A/G]GGTCCCAGGGGAGGCGGCCTTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606600 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASGRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASGRF1 - Ras protein specific guanine nucleotide releasing factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145648.1 | 5885 | UTR 3 | NP_001139120.1 | |||
NM_002891.4 | 5885 | UTR 3 | NP_002882.3 | |||
NM_153815.2 | 5885 | UTR 3 | NP_722522.1 | |||
XM_011521866.2 | 5885 | Intron | XP_011520168.1 | |||
XM_011521867.2 | 5885 | UTR 3 | XP_011520169.1 | |||
XM_017022455.1 | 5885 | UTR 3 | XP_016877944.1 | |||
XM_017022456.1 | 5885 | UTR 3 | XP_016877945.1 | |||
XM_017022457.1 | 5885 | Intron | XP_016877946.1 | |||
XM_017022458.1 | 5885 | Intron | XP_016877947.1 | |||
XM_017022459.1 | 5885 | UTR 3 | XP_016877948.1 |