Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCTCTCTCAATTTCAGCTTCG[C/G]AACGCTATGAAAATAATACATGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137028 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM227B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM227B - family with sequence similarity 227 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152647.2 | 1768 | Missense Mutation | TCC,TGC | S,C 475 | NP_689860.2 | |
XM_005254213.3 | 1768 | Missense Mutation | TCC,TGC | S,C 500 | XP_005254270.1 | |
XM_005254214.3 | 1768 | Missense Mutation | TCC,TGC | S,C 500 | XP_005254271.1 | |
XM_005254215.3 | 1768 | Missense Mutation | TCC,TGC | S,C 488 | XP_005254272.1 | |
XM_005254216.3 | 1768 | Missense Mutation | TCC,TGC | S,C 466 | XP_005254273.1 | |
XM_005254219.3 | 1768 | UTR 3 | XP_005254276.1 | |||
XM_005254220.3 | 1768 | Intron | XP_005254277.1 | |||
XM_006720423.2 | 1768 | Missense Mutation | TCC,TGC | S,C 500 | XP_006720486.1 | |
XM_006720424.2 | 1768 | Missense Mutation | TCC,TGC | S,C 475 | XP_006720487.1 | |
XM_006720426.2 | 1768 | Missense Mutation | TCC,TGC | S,C 453 | XP_006720489.1 | |
XM_006720427.3 | 1768 | Missense Mutation | TCC,TGC | S,C 417 | XP_006720490.1 | |
XM_011521319.2 | 1768 | Missense Mutation | TCC,TGC | S,C 500 | XP_011519621.1 | |
XM_011521320.1 | 1768 | Missense Mutation | TCC,TGC | S,C 366 | XP_011519622.1 | |
XM_011521321.2 | 1768 | Intron | XP_011519623.1 | |||
XM_011521322.1 | 1768 | Missense Mutation | TCC,TGC | S,C 354 | XP_011519624.1 | |
XM_011521324.1 | 1768 | Intron | XP_011519626.1 | |||
XM_011521325.2 | 1768 | Intron | XP_011519627.1 | |||
XM_011521326.1 | 1768 | Intron | XP_011519628.1 | |||
XM_017021990.1 | 1768 | Missense Mutation | TCC,TGC | S,C 419 | XP_016877479.1 | |
XM_017021991.1 | 1768 | Missense Mutation | TCC,TGC | S,C 417 | XP_016877480.1 | |
XM_017021992.1 | 1768 | Missense Mutation | TCC,TGC | S,C 400 | XP_016877481.1 | |
XM_017021993.1 | 1768 | Missense Mutation | TCC,TGC | S,C 394 | XP_016877482.1 | |
XM_017021994.1 | 1768 | Missense Mutation | TCC,TGC | S,C 386 | XP_016877483.1 | |
XM_017021995.1 | 1768 | UTR 3 | XP_016877484.1 | |||
XM_017021996.1 | 1768 | Missense Mutation | TCC,TGC | S,C 366 | XP_016877485.1 |
GALK2 - galactokinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001556.2 | 1768 | UTR 3 | NP_001001556.1 | |||
NM_001289030.1 | 1768 | UTR 3 | NP_001275959.1 | |||
NM_001289031.1 | 1768 | UTR 3 | NP_001275960.1 | |||
NM_002044.3 | 1768 | UTR 3 | NP_002035.1 | |||
XM_005254279.3 | 1768 | Intron | XP_005254336.1 | |||
XM_005254280.3 | 1768 | Intron | XP_005254337.1 | |||
XM_005254284.3 | 1768 | Intron | XP_005254341.1 | |||
XM_006720461.3 | 1768 | Missense Mutation | CAA,GAA | Q,E 452 | XP_006720524.1 | |
XM_006720462.3 | 1768 | Intron | XP_006720525.1 | |||
XM_006720463.3 | 1768 | Intron | XP_006720526.1 | |||
XM_006720464.3 | 1768 | Intron | XP_006720527.1 | |||
XM_011521441.2 | 1768 | Intron | XP_011519743.1 | |||
XM_017022062.1 | 1768 | UTR 3 | XP_016877551.1 | |||
XM_017022063.1 | 1768 | Intron | XP_016877552.1 | |||
XM_017022064.1 | 1768 | Intron | XP_016877553.1 | |||
XM_017022065.1 | 1768 | Missense Mutation | CAA,GAA | Q,E 323 | XP_016877554.1 | |
XM_017022066.1 | 1768 | Missense Mutation | CAA,GAA | Q,E 323 | XP_016877555.1 |