Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAACATGGACCTTGAAAGGGAT[A/G]CGCAGAGAATAGATGAAGCAAATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMCO5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMCO5A - transmembrane and coiled-coil domains 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152453.3 | 241 | Missense Mutation | ACG,GCG | T,A 24 | NP_689666.2 | |
XM_005254168.3 | 241 | Missense Mutation | ACG,GCG | T,A 24 | XP_005254225.1 | |
XM_011521262.2 | 241 | Missense Mutation | ACG,GCG | T,A 24 | XP_011519564.1 | |
XM_011521263.1 | 241 | Missense Mutation | ACG,GCG | T,A 24 | XP_011519565.1 | |
XM_011521264.1 | 241 | Missense Mutation | ACG,GCG | T,A 24 | XP_011519566.1 |