Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGAGAATTTCTTACATCATACT[A/G]GGAATTTTTCTTGTCTCTGTGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612188 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM87A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM87A - transmembrane protein 87A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110503.2 | Intron | NP_001103973.1 | ||||
NM_001286487.1 | Intron | NP_001273416.1 | ||||
NM_015497.4 | Intron | NP_056312.2 | ||||
XM_005254287.1 | Intron | XP_005254344.1 | ||||
XM_006720468.2 | Intron | XP_006720531.1 | ||||
XM_011521443.2 | Intron | XP_011519745.1 | ||||
XM_011521444.2 | Intron | XP_011519746.1 | ||||
XM_017022067.1 | Intron | XP_016877556.1 |
VPS39 - VPS39, HOPS complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |