Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTAGGCAAATGTTTGCAGACTAC[A/G]GCTGACCGTACCTCCTGAGAGTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614071 | ||||||||||||||||||||
Literature Links: |
GCOM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCOM1 - GRINL1A complex locus 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018090.6 | 226 | Missense Mutation | CAG,CGG | Q,R 32 | NP_001018100.1 | |
NM_001018091.6 | 226 | Missense Mutation | CAG,CGG | Q,R 32 | NP_001018101.1 | |
NM_001285900.3 | 226 | Missense Mutation | CAG,CGG | Q,R 32 | NP_001272829.1 |
MYZAP - myocardial zonula adherens protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018100.4 | 226 | Missense Mutation | CAG,CGG | Q,R 32 | NP_001018110.1 | |
NM_152451.7 | 226 | Missense Mutation | CAG,CGG | Q,R 32 | NP_689664.3 |