Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTTTCTTCACTTAACTGATCC[A/G]TAGAGGCATGACCATTTGATCGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603868 MIM: 613262 | ||||||||||||||||||||
Literature Links: |
RAB27A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB27A - RAB27A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004580.4 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | NP_004571.2 | |
NM_183234.2 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | NP_899057.1 | |
NM_183235.2 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | NP_899058.1 | |
NM_183236.2 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | NP_899059.1 | |
XM_005254576.4 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | XP_005254633.1 | |
XM_011521852.1 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | XP_011520154.1 | |
XM_011521854.1 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | XP_011520156.1 | |
XM_011521855.2 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | XP_011520157.1 | |
XM_011521856.2 | 1076 | Missense Mutation | ACG,ATG | T,M 207 | XP_011520158.1 |
RSL24D1 - ribosomal L24 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |