Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACATGTCCCTGCAGGACAGTGG[A/G]GGCCAGAGCCGGGCCATCAGCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190030 MIM: 136950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FES PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FES - FES proto-oncogene, tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143783.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | NP_001137255.1 | |
NM_001143784.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | NP_001137256.1 | |
NM_001143785.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | NP_001137257.1 | |
NM_002005.3 | 298 | Silent Mutation | GGA,GGG | G,G 57 | NP_001996.1 | |
XM_005254880.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | XP_005254937.1 | |
XM_005254882.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | XP_005254939.1 | |
XM_017022005.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | XP_016877494.1 | |
XM_017022006.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | XP_016877495.1 | |
XM_017022007.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | XP_016877496.1 | |
XM_017022008.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | XP_016877497.1 | |
XM_017022009.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | XP_016877498.1 | |
XM_017022010.1 | 298 | Silent Mutation | GGA,GGG | G,G 57 | XP_016877499.1 |
FURIN - furin, paired basic amino acid cleaving enzyme | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |