Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTTTGGCCGCCATGTGCTTCCC[A/G]AAGGTGAGTGAGAGGCTGCGTGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606001 MIM: 608482 | ||||||||||||||||||||
Literature Links: |
IL32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL32 - interleukin 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012631.1 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_001012649.1 | |
NM_001012632.1 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_001012650.1 | |
NM_001012633.1 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_001012651.1 | |
NM_001012634.1 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_001012652.1 | |
NM_001012635.1 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_001012653.1 | |
NM_001012636.1 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_001012654.1 | |
NM_001012718.1 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_001012736.1 | |
NM_001308078.1 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_001295007.1 | |
NM_004221.4 | 90 | Silent Mutation | CCA,CCG | P,P 4 | NP_004212.4 |
MMP25 - matrix metallopeptidase 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP25-AS1 - MMP25 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |