Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACGAGAGCACGTTGGACAGGGAC[C/G]CCGAGGCGTTCAGCTCCGAGGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612985 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRX3 - iroquois homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024336.2 | 862 | Missense Mutation | GCG,GGG | A,G 47 | NP_077312.2 | |
XM_005256139.3 | 862 | Missense Mutation | GCG,GGG | A,G 47 | XP_005256196.1 |