Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGATGCCACCGTTCTGAATGAC[A/G]GAGATCTCGTTGGCCTTCATCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602218 | ||||||||||||||||||||
Literature Links: |
LOC105371253 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105371253 - uncharacterized LOC105371253 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SALL1 - spalt like transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127892.1 | 3790 | Silent Mutation | TCC,TCT | S,S 1156 | NP_001121364.1 | |
NM_002968.2 | 3790 | Silent Mutation | TCC,TCT | S,S 1253 | NP_002959.2 | |
XM_006721241.3 | 3790 | Silent Mutation | TCC,TCT | S,S 1253 | XP_006721304.1 | |
XM_011523254.2 | 3790 | Silent Mutation | TCC,TCT | S,S 1253 | XP_011521556.1 |