Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCAAGGTGAAGCTGCCCATCAT[C/T]GGGGTGGTGGAGAACATGAGTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUBP1 - nucleotide binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278506.1 | 611 | Silent Mutation | ATC,ATT | I,I 213 | NP_001265435.1 | |
NM_001323594.1 | 611 | Silent Mutation | ATC,ATT | I,I 201 | NP_001310523.1 | |
NM_001323595.1 | 611 | Silent Mutation | ATC,ATT | I,I 224 | NP_001310524.1 | |
NM_001323596.1 | 611 | Silent Mutation | ATC,ATT | I,I 224 | NP_001310525.1 | |
NM_001323597.1 | 611 | Silent Mutation | ATC,ATT | I,I 175 | NP_001310526.1 | |
NM_002484.3 | 611 | Silent Mutation | ATC,ATT | I,I 224 | NP_002475.2 | |
XM_017023252.1 | 611 | Silent Mutation | ATC,ATT | I,I 201 | XP_016878741.1 |
TVP23A - trans-golgi network vesicle protein 23 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079512.3 | 611 | Intron | NP_001072980.1 | |||
NM_001318873.1 | 611 | Intron | NP_001305802.1 | |||
XM_006720944.3 | 611 | Intron | XP_006721007.1 | |||
XM_011522654.2 | 611 | Intron | XP_011520956.1 | |||
XM_017023649.1 | 611 | Intron | XP_016879138.1 | |||
XM_017023650.1 | 611 | Intron | XP_016879139.1 | |||
XM_017023651.1 | 611 | Intron | XP_016879140.1 |