Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGGAGCTTTCCTATGAGGCCT[A/G]CATGTCCCTCAGCATCACCAACAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114770 | ||||||||||||||||||||
Literature Links: |
CA7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CA7 - carbonic anhydrase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014435.1 | 270 | UTR 5 | NP_001014435.1 | |||
NM_005182.2 | 270 | Missense Mutation | TAC,TGC | Y,C 56 | NP_005173.1 | |
XM_005256136.3 | 270 | UTR 5 | XP_005256193.1 | |||
XM_011523311.1 | 270 | Missense Mutation | TAC,TGC | Y,C 56 | XP_011521613.1 | |
XM_011523312.1 | 270 | UTR 5 | XP_011521614.1 |