Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCCGACGAGCAGGGGCCTCTC[A/G]GGGGGCTTGGCTCACTGGGACAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611668 MIM: 614336 | ||||||||||||||||||||
Literature Links: |
CORO7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CORO7 - coronin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201472.1 | 2796 | Nonsense Mutation | CGA,TGA | R,* 852 | NP_001188401.1 | |
NM_001201473.1 | 2796 | Nonsense Mutation | CGA,TGA | R,* 785 | NP_001188402.1 | |
NM_024535.4 | 2796 | Nonsense Mutation | CGA,TGA | R,* 870 | NP_078811.3 |
CORO7-PAM16 - CORO7-PAM16 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201479.1 | 2796 | Nonsense Mutation | CGA,TGA | R,* 870 | NP_001188408.1 |
PAM16 - presequence translocase-associated motor 16 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |