Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTGCTGTCTCACAAAGCTGTC[A/T]GCAGACACTCGGAGTTTGGCTATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610609 MIM: 616886 | ||||||||||||||||||||
Literature Links: |
GINS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GINS2 - GINS complex subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSE1 - Gse1 coiled-coil protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |