Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGCACCTGTGAGCGCGAGTGTA[G/T]CAGGGACCAGGTGAGTGTGGTCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608021 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100287175 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100287175 - uncharacterized LOC100287175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL26 - methyltransferase like 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB40C - RAB40C, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WFIKKN1 - WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053284.2 | 403 | Missense Mutation | AGC,ATC | S,I 54 | NP_444514.1 |