Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTCCTCGCATGTTTTGCACTTA[C/T]GGGGCCTTTCCTCTGTATGAATGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614246 MIM: 614685 | ||||||||||||||||||||
Literature Links: |
NAA60 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAA60 - N(alpha)-acetyltransferase 60, NatF catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF597 - zinc finger protein 597 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152457.2 | 1337 | Missense Mutation | CAT,CGT | H,R 369 | NP_689670.1 |