Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGACACGGGATTGATCTATGACA[A/C]TCCCACTTCGGAATCTGGTCCCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602977 MIM: 191845 | ||||||||||||||||||||
Literature Links: |
GP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GP2 - glycoprotein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UMOD - uromodulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008389.2 | 2201 | Missense Mutation | AGT,ATT | S,I 591 | NP_001008390.1 | |
NM_001278614.1 | 2201 | Missense Mutation | AGT,ATT | S,I 624 | NP_001265543.1 | |
NM_003361.3 | 2201 | Missense Mutation | AGT,ATT | S,I 591 | NP_003352.2 | |
XM_011545934.1 | 2201 | Missense Mutation | AGT,ATT | S,I 666 | XP_011544236.1 | |
XM_011545935.1 | 2201 | Missense Mutation | AGT,ATT | S,I 638 | XP_011544237.1 | |
XM_011545936.1 | 2201 | Missense Mutation | AGT,ATT | S,I 638 | XP_011544238.1 | |
XM_011545937.1 | 2201 | Missense Mutation | AGT,ATT | S,I 638 | XP_011544239.1 | |
XM_011545938.1 | 2201 | Missense Mutation | AGT,ATT | S,I 638 | XP_011544240.1 | |
XM_011545940.1 | 2201 | Missense Mutation | AGT,ATT | S,I 687 | XP_011544242.1 | |
XM_017023641.1 | 2201 | Missense Mutation | AGT,ATT | S,I 619 | XP_016879130.1 |