Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGGGTCCCGGGATGCCCCTGCC[C/T]GAGCCCAGCGAGCAGGAGGGTGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FLYWCH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLYWCH2 - FLYWCH family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142499.1 | 289 | Silent Mutation | CCC,CCT | P,P 4 | NP_001135971.1 | |
NM_001142500.1 | 289 | Silent Mutation | CCC,CCT | P,P 4 | NP_001135972.1 | |
NM_138439.2 | 289 | Silent Mutation | CCC,CCT | P,P 4 | NP_612448.1 | |
XM_005255078.4 | 289 | Silent Mutation | CCC,CCT | P,P 4 | XP_005255135.1 |