Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCTGAGACAATATGAAGAAAGC[C/T]ACTCTATACATTAAGGTACAGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605812 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX19B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX19B - DEAD-box helicase 19B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014449.2 | Intron | NP_001014449.1 | ||||
NM_001014451.2 | Intron | NP_001014451.1 | ||||
NM_001257172.1 | Intron | NP_001244101.1 | ||||
NM_001257173.1 | Intron | NP_001244102.1 | ||||
NM_001257174.1 | Intron | NP_001244103.1 | ||||
NM_001257175.1 | Intron | NP_001244104.1 | ||||
NM_007242.5 | Intron | NP_009173.1 | ||||
XM_006721127.2 | Intron | XP_006721190.1 | ||||
XM_011522832.2 | Intron | XP_011521134.1 | ||||
XM_011522833.2 | Intron | XP_011521135.1 | ||||
XM_011522834.2 | Intron | XP_011521136.1 | ||||
XM_017022890.1 | Intron | XP_016878379.1 |